siRNA Details
Gene Symbol: CCDC78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522362.1 | XP_011520664.1 | 9 | 1970 | ||
GenBank mRNA | AK127495.1 | - | 4 | 2852 | ||
AK128538.1 | - | 4 | 3257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
