siRNA Details
Gene Symbol: AP2M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311198.1 | NP_001298127.1 | 6, 7 | 648 | ||
NM_004068.3 | NP_004059.2 | 4, 5 | 573 | |||
GenBank mRNA | AK057883.1 | - | 4, 5 | 564 | ||
AK294159.1 | - | 4, 5 | 394 | |||
AK295999.1 | - | 4, 5 | 357 | |||
AK297578.1 | - | 3, 4 | 248 | |||
AK297849.1 | - | 6, 7 | 639 | |||
BC004996.1 | - | 4, 5 | 551 | 2958005 | ||
BC014030.1 | - | 4, 5 | 541 | 4541249 | ||
BT007308.1 | - | 3, 4 | 416 | |||
D63475.1 | - | 4, 5 | 502 | |||
U36188.1 | - | 3, 4 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
