siRNA Details
Gene Symbol: C12orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097445.1 | - | 19 | 2498 | ||
EU668334.1 | - | 19 | 2498 | |||
GQ472216.1 | - | 19 | 2498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
