siRNA Details
Gene Symbol: DPY19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173812.4 | NP_776173.3 | 3 | 613 | ||
XM_006719352.2 | XP_006719415.1 | 3 | 539 | |||
XM_017019192.1 | XP_016874681.1 | 3 | 648 | |||
XM_017019193.1 | XP_016874682.1 | 2 | 195 | |||
XM_017019195.1 | XP_016874684.1 | 3 | 270 | |||
XM_017019201.1 | XP_016874690.1 | 3 | 548 | |||
XM_017019203.1 | XP_016874692.1 | 3 | 443 | |||
XM_017019204.1 | XP_016874693.1 | 4 | 681 | |||
GenBank mRNA | AK057511.1 | - | 3 | 584 | ||
AL833344.1 | - | 3 | 540 | |||
AY358792.1 | - | 3 | 534 | |||
BC068442.1 | - | 3 | 616 | 5297524 | ||
BC125215.1 | - | 3 | 503 | |||
BC125216.1 | - | 3 | 503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
