siRNA Details
Gene Symbol: PIP4K2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003559.4 | NP_003550.1 | 10 | 3631 | ||
XM_011525326.2 | XP_011523628.1 | 11 | 3772 | |||
XM_011525327.2 | XP_011523629.1 | 12 | 3291 | |||
XM_011525330.2 | XP_011523632.1 | 9 | 2967 | |||
XM_017025196.1 | XP_016880685.1 | 11 | 3208 | |||
XM_017025197.1 | XP_016880686.1 | 11 | 3108 | |||
XM_017025198.1 | XP_016880687.1 | 9 | 3594 | |||
XM_017025199.1 | XP_016880688.1 | 10 | 3046 | |||
GenBank mRNA | U85245.1 | - | 10 | 3626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
