siRNA Details
Gene Symbol: BOD1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257964.1 | NP_001244893.1 | 1 | 541 | ||
GenBank mRNA | AK127964.1 | - | 1 | 541 | ||
AK301799.1 | - | 2 | 363 | |||
BC021740.1 | - | 1 | 505 | |||
BC035018.1 | - | 1 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
