siRNA Details
Gene Symbol: TMEM105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178520.3 | NP_848615.1 | 3 | 1179 | ||
XM_017024522.1 | XP_016880011.1 | 3 | 1113 | |||
GenBank mRNA | AK096111.1 | - | 3 | 1179 |
