siRNA Details
Gene Symbol: C1QTNF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007537.2 | NP_001007538.1 | 3 | 599 | ||
XM_011535070.2 | XP_011533372.1 | 4 | 1729 | |||
GenBank mRNA | BC137004.1 | - | 3 | 648 | 9020997 | |
BC137006.1 | - | 3 | 648 | 9020999 |
Gene Symbol: C1QTNF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303137.1 | NP_001290066.1 | 5 | 724 | ||
NM_001303138.1 | NP_001290067.1 | 4 | 664 | |||
NM_178540.4 | NP_848635.2 | 4 | 640 | |||
GenBank mRNA | AY358145.1 | - | 4 | 635 | ||
BC040438.1 | - | 4 | 640 | 5227612 | ||
BI767787.1 | - | 4 | 661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
