siRNA Details
Gene Symbol: C1QTNF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303137.1 | NP_001290066.1 | 5 | 1110 | ||
NM_001303138.1 | NP_001290067.1 | 4 | 1050 | |||
NM_178540.4 | NP_848635.2 | 4 | 1026 | |||
GenBank mRNA | AY358145.1 | - | 4 | 1021 | ||
BC040438.1 | - | 4 | 1026 | 5227612 |
Gene Symbol: C1QTNF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007537.2 | NP_001007538.1 | 3 | 985 | ||
NR_104426.1 | 6 | 661 | ||||
XM_011535070.2 | XP_011533372.1 | 4 | 2115 | |||
GenBank mRNA | BC110413.1 | - | 6 | 661 | ||
BC137004.1 | - | 3 | 1034 | 9020997 | ||
BC137006.1 | - | 3 | 1034 | 9020999 | ||
BC144450.1 | - | 5 | 650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
