siRNA Details
Gene Symbol: TMTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181783.3 | NP_861448.2 | 10, 11 | 1656 | ||
XM_011537980.2 | XP_011536282.1 | 9, 10 | 1431 | |||
XM_011537981.2 | XP_011536283.1 | 3, 4 | 320 | |||
GenBank mRNA | AJ697717.1 | - | 10, 11 | 1526 | ||
AK074973.1 | - | 9, 10 | 1283 | |||
AK290436.1 | - | 10, 11 | 1656 | |||
AK302755.1 | - | 10, 11 | 1656 | |||
AK310375.1 | - | 11, 12 | 1780 | |||
BC117175.1 | - | 9, 10 | 1477 | 40125726 | ||
BC117177.1 | - | 9, 10 | 1478 | 40125728 | ||
BX538169.1 | - | 10, 11 | 1629 | |||
CR749244.1 | - | 10, 11 | 1608 | |||
CR749309.1 | - | 10, 11 | 1642 |
