siRNA Details
Gene Symbol: GOLGA6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304388.1 | NP_001291317.1 | 8 | 3033 | ||
GenBank mRNA | AK093463.1 | - | 9 | 2054 | ||
AK097414.1 | - | 1760 | ||||
AK128822.1 | - | 1722 |
