siRNA Details
Gene Symbol: GOLGA6L22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271664.2 | NP_001258593.2 | 10 | 2697 |
Gene Symbol: GOLGA6L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001413.3 | NP_001001413.3 | 9 | 2713 |
Gene Symbol: GOLGA6L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145004.2 | NP_001138476.2 | 9 | 2904 | ||
GenBank mRNA | AK093450.1 | - | 2 | 869 |
Gene Symbol: LOC102723623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720795.3 | XP_006720858.1 | 9 | 2732 | ||
XM_017022778.1 | XP_016878267.1 | 10 | 2805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
