siRNA Details
Gene Symbol: B3GNTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126018.1 | - | 1 | 2719 | ||
BC110528.1 | - | 1 | 2299 | 40034264 | ||
BC110529.1 | - | 1 | 2299 | 40034267 | ||
U79265.1 | - | 1 | 1139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
