siRNA Details
Gene Symbol: C12orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099336.2 | NP_001092806.1 | 5 | 667 | ||
NM_001278419.1 | NP_001265348.1 | 6 | 762 | |||
NM_001278420.1 | NP_001265349.1 | 6 | 627 | |||
NM_198521.3 | NP_940923.2 | 5 | 532 | |||
XM_006719377.3 | XP_006719440.1 | 4 | 470 | |||
XM_011538291.1 | XP_011536593.1 | 5 | 527 | |||
XM_011538292.2 | XP_011536594.1 | 5 | 496 | |||
XM_011538293.2 | XP_011536595.1 | 5 | 503 | |||
XM_011538294.2 | XP_011536596.1 | 5 | 477 | |||
XM_011538295.1 | XP_011536597.1 | 5 | 559 | |||
XM_011538297.2 | XP_011536599.1 | 6 | 1543 | |||
XM_011538300.1 | XP_011536602.1 | 6 | 712 | |||
XM_011538305.1 | XP_011536607.1 | 5 | 729 | |||
XM_017019276.1 | XP_016874765.1 | 5 | 621 | |||
XM_017019277.1 | XP_016874766.1 | 8 | 996 | |||
XM_017019278.1 | XP_016874767.1 | 7 | 775 | |||
XM_017019279.1 | XP_016874768.1 | 8 | 1301 | |||
XM_017019280.1 | XP_016874769.1 | 8 | 1001 | |||
GenBank mRNA | AK058052.1 | - | 5 | 490 | ||
BC044617.1 | - | 6 | 723 | 5267756 | ||
BC117392.1 | - | 5 | 490 | 40125943 | ||
BC117394.1 | - | 5 | 490 | 40125945 | ||
BI830750.1 | - | 5 | 610 |
