siRNA Details
Gene Symbol: EFCAB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198529.3 | NP_940931.2 | 23 | 4615 | ||
XM_011524759.1 | XP_011523061.1 | 23 | 4621 | |||
XM_011524765.1 | XP_011523067.1 | 20 | 3983 | |||
XM_011524766.2 | XP_011523068.1 | 18 | 3561 | |||
XM_017024591.1 | XP_016880080.1 | 20 | 3700 | |||
GenBank mRNA | AK128126.1 | - | 18 | 3469 | ||
AL833911.1 | - | 4 | 657 | |||
BC110608.1 | - | 20 | 3818 | |||
BC110609.1 | - | 20 | 3818 | |||
BC119807.1 | - | 12 | 2234 | |||
BC137519.1 | - | 23 | 4451 | 9021519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
