siRNA Details
Gene Symbol: CDK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003718.4 | NP_003709.3 | 5, 6 | 2643 | ||
NM_031267.3 | NP_112557.2 | 5, 6 | 2643 | |||
XM_011515597.2 | XP_011513899.1 | 5, 6 | 3502 | |||
XM_017012750.1 | XP_016868239.1 | 5, 6 | 3502 | |||
XM_017012751.1 | XP_016868240.1 | 5, 6 | 3502 | |||
GenBank mRNA | AJ297709.1 | - | 5, 6 | 2493 | ||
AJ297710.1 | - | 5, 6 | 2493 | |||
BC001274.1 | - | 4, 5 | 913 | |||
M80629.1 | - | 5, 6 | 1290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
