siRNA Details
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001016.3 | NP_001007.2 | 1 | 315 | ||
GenBank mRNA | AK311826.1 | - | 4, 5 | 310 | ||
BC002550.1 | - | 1400 | ||||
BC017321.2 | - | 1 | 298 | 4565075 | ||
BC071930.1 | - | 1 | 294 | 6295016 | ||
BC095424.1 | - | 1 | 305 | 30792687 | ||
BQ276878.1 | - | 4, 5 | 318 | |||
X53505.1 | - | 4, 5 | 312 |
Gene Symbol: PUS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011532568.2 | XP_011530870.1 | 19 | 5961 | ||
XM_011532570.1 | XP_011530872.1 | 18 | 5668 | |||
XM_011532571.1 | XP_011530873.1 | 18 | 5842 | |||
XM_011532572.1 | XP_011530874.1 | 18 | 5762 | |||
XM_011532573.2 | XP_011530875.1 | 18 | 5722 | |||
XM_011532574.2 | XP_011530876.1 | 18 | 5783 | |||
XM_011532576.2 | XP_011530878.1 | 18 | 5842 | |||
XM_011532578.1 | XP_011530880.1 | 18 | 5820 | |||
XM_017003428.1 | XP_016858917.1 | 17 | 5659 | |||
XM_017003429.1 | XP_016858918.1 | 17 | 5455 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001037166, BC100581.
Gene Ontology Categories:
