siRNA Details
Gene Symbol: PRKCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002738.6 | NP_002729.2 | 17 | 2223 | ||
GenBank mRNA | BC036472.1 | - | 17 | 2183 | 5260547 | |
M13975.1 | - | 17 | 2090 | |||
X07109.1 | - | 17 | 2162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
