siRNA Details
Gene Symbol: WARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004184.3 | NP_004175.2 | 2, 3 | 488 | ||
NM_173701.1 | NP_776049.1 | 2, 3 | 264 | |||
XM_005268044.3 | XP_005268101.1 | 2, 3 | 216 | |||
XM_006720249.3 | XP_006720312.1 | 3, 4 | 450 | |||
XM_011537133.2 | XP_011535435.1 | 4, 5 | 462 | |||
XM_011537135.2 | XP_011535437.1 | 3, 4 | 346 | |||
XM_011537136.2 | XP_011535438.1 | 2, 3 | 182 | |||
XM_017021627.1 | XP_016877116.1 | 3, 4 | 354 | |||
XM_017021628.1 | XP_016877117.1 | 3, 4 | 485 | |||
XM_017021629.1 | XP_016877118.1 | 4, 5 | 460 | |||
GenBank mRNA | AK056100.1 | - | 2, 3 | 726 | ||
AK300255.1 | - | 2, 3 | 371 | |||
BC017489.1 | - | 2, 3 | 364 | 3542671 | ||
BC095453.1 | - | 3, 4 | 375 | 30418785 | ||
BM834473.1 | - | 3, 4 | 600 | |||
BX248006.1 | - | 2, 3 | 208 | |||
M61715.1 | - | 2, 3 | 294 | |||
M77804.1 | - | 1, 2 | 149 | |||
X59892.1 | - | 2, 3 | 218 | |||
X62570.1 | - | 1, 2 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
