siRNA Details
Gene Symbol: QSOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002826.4 | NP_002817.2 | 12 | 2454 | ||
GenBank mRNA | AY358941.1 | - | 12 | 2405 | ||
BC100023.1 | - | 12 | 2398 | 30563992 | ||
L42379.1 | - | 12 | 2377 | |||
U97276.2 | - | 12 | 2454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
