siRNA Details
Gene Symbol: ITSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003024.2 | NP_003015.2 | 30 | 3987 | ||
XM_005261026.2 | XP_005261083.1 | 29 | 4050 | |||
XM_017028428.1 | XP_016883917.1 | 31 | 3985 | |||
XM_017028429.1 | XP_016883918.1 | 30 | 3954 | |||
XM_017028430.1 | XP_016883919.1 | 29 | 3939 | |||
XM_017028431.1 | XP_016883920.1 | 30 | 4065 | |||
XM_017028432.1 | XP_016883921.1 | 29 | 4050 | |||
XM_017028433.1 | XP_016883922.1 | 28 | 3852 | |||
XM_017028434.1 | XP_016883923.1 | 27 | 3837 | |||
XM_017028435.1 | XP_016883924.1 | 28 | 3741 | |||
XM_017028436.1 | XP_016883925.1 | 27 | 3726 | |||
XM_017028437.1 | XP_016883926.1 | 25 | 3405 | |||
GenBank mRNA | AF064244.1 | - | 30 | 3825 | ||
AF114487.1 | - | 30 | 3987 | |||
BC116185.1 | - | 29 | 3783 | 40073780 | ||
BC116186.2 | - | 30 | 3798 | 40073781 | ||
EU117382.1 | - | 27 | 3538 | |||
EU120733.1 | - | 28 | 3705 | |||
EU120734.1 | - | 28 | 3705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
