siRNA Details
Gene Symbol: ACVR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004302.4 | NP_004293.1 | 8 | 1408 | ||
NM_020327.3 | NP_064732.3 | 8 | 1360 | |||
NM_020328.3 | NP_064733.3 | 9 | 1531 | |||
XM_011538966.2 | XP_011537268.1 | 8 | 1384 | |||
XM_017020199.1 | XP_016875688.1 | 9 | 2229 | |||
XM_017020200.1 | XP_016875689.1 | 10 | 1624 | |||
XM_017020201.1 | XP_016875690.1 | 7 | 1261 | |||
GenBank mRNA | AK299120.1 | - | 9 | 1494 | ||
AK299496.1 | - | 8 | 1360 | |||
BC000254.1 | - | 8 | 1357 | 3352720 | ||
BC040531.1 | - | 8 | 1405 | 5260767 | ||
BT007072.1 | - | 8 | 1331 | |||
L10125.1 | - | 8 | 1370 | |||
U14722.1 | - | 8 | 1331 | |||
Z22536.1 | - | 8 | 1331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
