siRNA Details
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270526.1 | NP_001257455.1 | 5 | 458 | ||
NM_001270527.1 | NP_001257456.1 | 5 | 458 | |||
NM_004099.5 | NP_004090.4 | 5 | 458 | |||
NR_073037.1 | 6 | 588 | ||||
GenBank mRNA | AK302977.1 | - | 5 | 384 | ||
AK304449.1 | - | 6 | 532 | |||
AK304518.1 | - | 5 | 402 | |||
AK307999.1 | - | 5 | 402 | |||
BC010703.1 | - | 5 | 380 | 3873494 | ||
BI553227.1 | - | 5 | 387 | |||
BM451470.1 | - | 422 | ||||
BP208523.1 | - | 5 | 385 | |||
CR542100.1 | - | 5 | 341 | |||
DC368197.1 | - | 5 | 458 | |||
M81635.1 | - | 5 | 362 | |||
X60067.1 | - | 5 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
