siRNA Details
Gene Symbol: SRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146282.1 | NP_001139754.1 | 13 | 1492 | ||
NM_003136.3 | NP_003127.1 | 14 | 1584 | |||
XM_011537106.1 | XP_011535408.1 | 14 | 1465 | |||
XM_017021615.1 | XP_016877104.1 | 13 | 1424 | |||
GenBank mRNA | AK293319.1 | - | 14 | 1376 | ||
AK300824.1 | - | 13 | 1365 | |||
AK312853.1 | - | 14 | 1529 | |||
BC000652.2 | - | 14 | 1451 | 3343314 | ||
BC003389.1 | - | 14 | 1457 | 3446599 | ||
U51920.1 | - | 14 | 1304 |
