siRNA Details
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 16 | 3522 | ||
GenBank mRNA | AB209991.1 | - | 16 | 3271 | ||
AK303241.1 | - | 16 | 1956 | |||
BC066122.1 | - | 16 | 3353 | |||
X02596.1 | - | 16 | 3414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
