siRNA Details
Gene Symbol: TAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286074.1 | NP_001273003.1 | 38 | 5462 | ||
NM_004606.4 | NP_004597.2 | 37 | 5456 | |||
NM_138923.3 | NP_620278.1 | 37 | 5393 | |||
NR_104387.1 | 38 | 5558 | ||||
NR_104388.1 | 38 | 5549 | ||||
NR_104389.1 | 37 | 5456 | ||||
NR_104390.1 | 38 | 5462 | ||||
NR_104391.1 | 39 | 5484 | ||||
NR_104392.1 | 38 | 5478 | ||||
NR_104393.1 | 39 | 5564 | ||||
NR_104394.1 | 38 | 5435 | ||||
NR_104395.1 | 38 | 5487 | ||||
XM_005262295.1 | XP_005262352.1 | 38 | 5492 | |||
XM_005262296.1 | XP_005262353.1 | 38 | 5489 | |||
XM_005262297.4 | XP_005262354.1 | 38 | 5514 | |||
XM_006724682.3 | XP_006724745.1 | 37 | 6436 | |||
GenBank mRNA | AB209316.1 | - | 23 | 2913 | ||
AB300418.1 | - | 38 | 5389 | |||
AJ549247.1 | - | 2 | 138 | |||
AJ549248.3 | - | 13 | 1383 | |||
AJ549249.2 | - | 9 | 920 | |||
AJ549250.3 | - | 12 | 1281 | |||
AJ555148.2 | - | 13 | 1287 | |||
AJ555149.2 | - | 14 | 1309 | |||
AM711892.1 | - | 13 | 1303 | |||
AM711893.1 | - | 14 | 1389 | |||
AM711894.1 | - | 13 | 1260 | |||
AM711895.1 | - | 13 | 1312 | |||
D90359.1 | - | 37 | 5368 | |||
M73444.1 | - | 7 | 741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
