siRNA Details
Gene Symbol: RXRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256570.1 | NP_001243499.1 | 6 | 1197 | ||
NM_001256571.1 | NP_001243500.1 | 4 | 614 | |||
NM_006917.4 | NP_008848.1 | 5 | 1139 | |||
GenBank mRNA | AB307706.1 | - | 5 | 677 | ||
AB593015.1 | - | 5 | 783 | |||
AB593016.1 | - | 6 | 1105 | |||
AK312922.1 | - | 5 | 1069 | |||
BC012063.1 | - | 5 | 865 | 4635470 | ||
BI768747.1 | - | 4 | 568 | |||
CR456705.1 | - | 5 | 677 | |||
HQ692845.1 | - | 5 | 677 | |||
U38480.1 | - | 5 | 704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
