siRNA Details
Gene Symbol: SUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005849.1 | NP_001005849.1 | 2 | 292 | ||
NM_006937.3 | NP_008868.3 | 2 | 292 | |||
GenBank mRNA | AK024823.1 | - | 2 | 193 | ||
AK225090.1 | - | 2 | 203 | |||
AK225263.1 | - | 2 | 204 | |||
AK311837.1 | - | 2 | 238 | |||
BC008450.1 | - | 2 | 259 | 4133768 | ||
BC016775.1 | - | 2 | 273 | 4096440 | ||
BC022340.1 | - | 2 | 243 | 4134531 | ||
BC062713.1 | - | 2 | 206 | 30404344 | ||
BC068465.1 | - | 2 | 258 | 5277638 | ||
BC070159.1 | - | 2 | 266 | 6592382 | ||
BC071645.1 | - | 2 | 199 | 3884132 | ||
BC071646.1 | - | 2 | 235 | 4369354 | ||
BC107853.1 | - | 2 | 196 | 5772748 | ||
BF700114.1 | - | 2 | 259 | |||
BG778261.1 | - | 2 | 293 | |||
L76416.1 | - | 2 | 137 | |||
X99585.1 | - | 2 | 213 |
Gene Symbol: SUMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002255.1 | NP_001002255.1 | 1 | 156 | ||
GenBank mRNA | AY340238.1 | - | 1 | 156 | ||
BC130305.1 | - | 1 | 150 | 40146336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
