siRNA Details
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 5 | 346 | ||
NM_001012632.1 | NP_001012650.1 | 4 | 145 | |||
NM_001012633.1 | NP_001012651.1 | 4 | 279 | |||
NM_001012636.1 | NP_001012654.1 | 3 | 96 | |||
NM_001012718.1 | NP_001012736.1 | 4 | 282 | |||
NM_001308078.1 | NP_001295007.1 | 3 | 280 | |||
NM_004221.4 | NP_004212.4 | 4 | 279 | |||
GenBank mRNA | AK307158.1 | - | 2 | 663 | ||
AK310053.1 | - | 3 | 283 | |||
AY495332.1 | - | 4 | 269 | |||
AY495333.1 | - | 3 | 227 | |||
BC009401.2 | - | 3 | 417 | 4300407 | ||
BC018782.1 | - | 4 | 126 | 4873608 | ||
BC105602.1 | - | 4 | 147 | 6496101 | ||
BF568456.1 | - | 3 | 418 | |||
BG748458.1 | - | 4 | 225 | |||
BM759359.1 | - | 1 | 20 | |||
BM822755.1 | - | 5 | 254 | |||
FJ985780.1 | - | 4 | 233 | |||
M59807.1 | - | 3 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
