siRNA Details
Gene Symbol: MBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025081.1 | NP_001020252.1 | 6 | 621 | ||
NM_001025090.1 | NP_001020261.1 | 5 | 543 | |||
NM_001025092.1 | NP_001020263.1 | 4 | 510 | |||
NM_001025101.1 | NP_001020272.1 | 8 | 1115 | |||
NM_002385.2 | NP_002376.1 | 5 | 588 | |||
GenBank mRNA | AK098402.1 | - | 5 | 499 | ||
AK122594.1 | - | 1 | 1066 | |||
AK123433.1 | - | 1 | 3163 | |||
AK124830.1 | - | 1 | 1840 | |||
AK128770.1 | - | 5 | 502 | |||
AK128788.1 | - | 5 | 499 | |||
AK289893.1 | - | 5 | 543 | |||
AK293922.1 | - | 4 | 538 | |||
AK314553.1 | - | 6 | 617 | |||
BC008749.2 | - | 5 | 481 | 3346521 | ||
BC068550.1 | - | 3 | 357 | |||
BC080654.1 | - | 4 | 455 | 4859683 | ||
BC101771.1 | - | 5 | 481 | 8069277 | ||
BC101773.1 | - | 5 | 481 | 8069279 | ||
BC143348.1 | - | 4 | 448 | 9051853 | ||
BC143350.1 | - | 5 | 481 | 9051855 | ||
CB153535.1 | - | 6 | 508 | |||
CB156561.1 | - | 5 | 416 | |||
CR536534.1 | - | 4 | 419 | |||
CR541919.1 | - | 4 | 419 | |||
CR627018.1 | - | 4 | 3970 | |||
M13577.1 | - | 5 | 488 | |||
M30047.1 | - | 4 | 451 | |||
M30515.1 | - | 6 | 537 | |||
M30516.1 | - | 5 | 507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
