siRNA Details
Gene Symbol: MKI67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145966.1 | NP_001139438.1 | 12 | 2121 | ||
NM_002417.4 | NP_002408.3 | 13 | 3201 | |||
XM_006717864.3 | XP_006717927.2 | 2 | 1102 | |||
XM_011539818.2 | XP_011538120.1 | 10 | 2932 | |||
GenBank mRNA | X65550.1 | - | 13 | 3202 | ||
X65551.1 | - | 12 | 2122 |
