siRNA Details
Gene Symbol: MSH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289856.1 | - | 2, 3 | 291 | ||
BC004177.2 | - | 2, 3 | 263 | |||
BC011817.2 | - | 2, 3 | 263 | |||
BC017273.2 | - | 1, 2 | 245 | |||
BC098436.1 | - | 2, 3 | 263 | |||
J04810.1 | - | 1, 2 | 303 | |||
U61981.1 | - | 2, 3 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
