siRNA Details
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 6 | 1503 | ||
NM_001197317.2 | NP_001184246.1 | 4 | 1310 | |||
NM_001197318.2 | NP_001184247.1 | 4 | 1317 | |||
NM_001197319.2 | NP_001184248.1 | 3 | 1206 | |||
NM_013269.5 | NP_037401.1 | 5 | 1421 | |||
NR_036693.2 | 4 | 1392 | ||||
GenBank mRNA | AK310554.1 | - | 4 | 1383 | ||
BC094859.1 | - | 1 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
