siRNA Details
Gene Symbol: PDPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261816.1 | NP_001248745.1 | 4 | 502 | ||
NM_002613.4 | NP_002604.1 | 4 | 502 | |||
XM_011522521.2 | XP_011520823.1 | 4 | 4190 | |||
XM_017023271.1 | XP_016878760.1 | 4 | 4195 | |||
XM_017023272.1 | XP_016878761.1 | 4 | 751 | |||
XM_017023273.1 | XP_016878762.1 | 4 | 460 | |||
XM_017023274.1 | XP_016878763.1 | 5 | 598 | |||
GenBank mRNA | AB209835.1 | - | 4 | 469 | ||
AF017995.1 | - | 4 | 433 | |||
AJ785968.1 | - | 3 | 1136 | |||
AK131060.1 | - | 4 | 436 | |||
AK222581.1 | - | 4 | 445 | |||
AK293900.1 | - | 3 | 693 | |||
BC012103.1 | - | 4 | 462 | 4561371 | ||
BI830733.1 | - | 4 | 499 | |||
CR536517.1 | - | 4 | 353 | |||
Y15056.1 | - | 4 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
