siRNA Details
Gene Symbol: PGAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317079.1 | NP_001304008.1 | 2 | 408 | ||
NM_002629.3 | NP_002620.1 | 2 | 478 | |||
GenBank mRNA | AK223096.1 | - | 2 | 324 | ||
AK292216.1 | - | 2 | 379 | |||
AK293023.1 | - | 2 | 323 | |||
AK296619.1 | - | 2 | 267 | |||
AK312351.1 | - | 2 | 307 | |||
AK315910.1 | - | 2 | 411 | |||
AY007118.1 | - | 1 | 315 | |||
BC000455.1 | - | 1 | 341 | 2821567 | ||
BC010038.1 | - | 1 | 333 | 3604026 | ||
BC011678.2 | - | 1 | 297 | 4299008 | ||
BC053356.1 | - | 1 | 314 | 5787678 | ||
BC062302.1 | - | 1 | 313 | 6584466 | ||
BC066959.1 | - | 2 | 346 | 5262935 | ||
BC073742.1 | - | 1 | 312 | 6297173 | ||
BC157873.1 | - | 2 | 330 | 8862699 | ||
CR541659.1 | - | 2 | 295 | |||
CR541744.1 | - | 2 | 295 | |||
EU794644.1 | - | 2 | 307 | |||
J04173.1 | - | 2 | 326 |
Gene Symbol: PGAM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029891.2 | NP_001025062.1 | 1 | 295 | ||
GenBank mRNA | BG875904.1 | - | 1 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
