siRNA Details
Gene Symbol: PIGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002643.3 | NP_002634.1 | 6 | 921 | ||
NM_173074.2 | NP_775097.1 | 7 | 1026 | |||
XM_011532908.2 | XP_011531210.1 | 7 | 908 | |||
GenBank mRNA | AK291108.1 | - | 6 | 876 | ||
BC021725.1 | - | 7 | 966 | 5269689 | ||
BC029408.1 | - | 6 | 874 | 4664848 | ||
D13435.1 | - | 6 | 824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
