siRNA Details
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031803.1 | NP_001026973.1 | 26 | 3254 | ||
NM_004524.2 | NP_004515.2 | 25 | 3211 | |||
XM_011524801.1 | XP_011523103.1 | 26 | 3249 | |||
XM_011524802.1 | XP_011523104.1 | 26 | 3237 | |||
XM_011524807.2 | XP_011523109.1 | 26 | 3354 | |||
XM_017024625.1 | XP_016880114.1 | 26 | 3234 | |||
XM_017024626.1 | XP_016880115.1 | 26 | 3225 | |||
XM_017024627.1 | XP_016880116.1 | 26 | 3222 | |||
XM_017024628.1 | XP_016880117.1 | 25 | 3194 | |||
XM_017024629.1 | XP_016880118.1 | 25 | 3191 | |||
XM_017024630.1 | XP_016880119.1 | 25 | 3182 | |||
XM_017024632.1 | XP_016880121.1 | 16 | 2606 | |||
GenBank mRNA | AK025401.1 | - | 15 | 2679 | ||
AK126687.1 | - | 22 | 3234 | |||
AK301204.1 | - | 18 | 2088 | |||
BC031842.1 | - | 9 | 853 | |||
BC064994.1 | - | 26 | 3227 | 5575350 | ||
X87342.1 | - | 25 | 3187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
