siRNA Details
Gene Symbol: LIMK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005569.3 | NP_005560.1 | 15, 16 | 1922 | ||
NM_016733.2 | NP_057952.1 | 14, 15 | 2069 | |||
GenBank mRNA | AB451269.1 | - | 15, 16 | 1777 | ||
AB451396.1 | - | 15, 16 | 1777 | |||
AK093554.1 | - | 10, 11 | 1169 | |||
AK291640.1 | - | 15, 16 | 1891 | |||
AK293774.1 | - | 15, 16 | 1992 | |||
AK299112.1 | - | 14, 15 | 2094 | |||
AK302553.1 | - | 15, 16 | 1828 | |||
AK314391.1 | - | 14, 15 | 2047 | |||
AL117466.1 | - | 14, 15 | 2029 | |||
CR456513.1 | - | 15, 16 | 1810 | |||
D45906.1 | - | 15, 16 | 1891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
