siRNA Details
Gene Symbol: LIMK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005569.3 | NP_005560.1 | 16 | 2743 | ||
NM_016733.2 | NP_057952.1 | 15 | 2890 | |||
GenBank mRNA | AK092838.1 | - | 1 | 1264 | ||
AK093554.1 | - | 12 | 1910 | |||
AL117466.1 | - | 15 | 2850 | |||
D45906.1 | - | 16 | 2712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
