siRNA Details
Gene Symbol: MFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005926.2 | NP_005917.2 | 9 | 1351 | ||
GenBank mRNA | AK312867.1 | - | 9 | 1327 | ||
BC023557.2 | - | 9 | 1286 | 4139958 | ||
BC050742.1 | - | 9 | 1297 | 6200464 | ||
U04209.1 | - | 9 | 1285 |
