siRNA Details
Gene Symbol: NOP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258308.1 | NP_001245237.1 | 4, 5 | 366 | ||
NM_001258309.1 | NP_001245238.1 | 4, 5 | 366 | |||
XM_005253691.1 | XP_005253748.1 | 4, 5 | 399 | |||
GenBank mRNA | AK307582.1 | - | 4, 5 | 348 | ||
AU131299.1 | - | 4, 5 | 348 | |||
BC000656.2 | - | 4, 5 | 287 | 3349415 | ||
BC106072.1 | - | 4, 5 | 312 | 6169852 | ||
BC128183.1 | - | 4, 5 | 341 | 40113364 | ||
BC128184.1 | - | 4, 5 | 341 | 40113367 | ||
DC316656.1 | - | 4, 5 | 366 | |||
M32110.1 | - | 3, 4 | 280 | |||
X55504.1 | - | 3, 4 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
