siRNA Details
Gene Symbol: NTRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018064.2 | NP_001018074.1 | 17 | 3206 | ||
NM_006180.4 | NP_006171.2 | 21 | 3913 | |||
XM_005252001.2 | XP_005252058.1 | 20 | 3794 | |||
XM_005252003.2 | XP_005252060.1 | 20 | 3610 | |||
XM_005252004.2 | XP_005252061.1 | 19 | 3510 | |||
XM_011518718.2 | XP_011517020.1 | 19 | 3746 | |||
XM_017014751.1 | XP_016870240.1 | 21 | 4336 | |||
XM_017014752.1 | XP_016870241.1 | 20 | 3801 | |||
XM_017014753.1 | XP_016870242.1 | 20 | 4288 | |||
XM_017014754.1 | XP_016870243.1 | 18 | 3462 | |||
GenBank mRNA | AF410899.1 | - | 19 | 3365 | ||
S76473.1 | - | 17 | 2948 | |||
U12140.1 | - | 17 | 3186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
