siRNA Details
Gene Symbol: SLC28A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK291997.1 | - | 7 | 781 | ||
U62967.1 | - | 5 | 573 | |||
U62968.1 | - | 5 | 573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
