siRNA Details
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 8 | 607 | ||
NM_001012632.1 | NP_001012650.1 | 7 | 406 | |||
NM_001012634.1 | NP_001012652.1 | 6 | 347 | |||
NM_001012635.1 | NP_001012653.1 | 6 | 342 | |||
NM_001012636.1 | NP_001012654.1 | 5 | 330 | |||
NM_001012718.1 | NP_001012736.1 | 7 | 543 | |||
NM_001308078.1 | NP_001295007.1 | 6 | 541 | |||
NM_004221.4 | NP_004212.4 | 7 | 540 | |||
GenBank mRNA | AK296143.1 | - | 6 | 352 | ||
AY495332.1 | - | 7 | 530 | |||
AY495333.1 | - | 6 | 488 | |||
BC009401.2 | - | 6 | 678 | 4300407 | ||
BC018782.1 | - | 7 | 387 | 4873608 | ||
BC105602.1 | - | 7 | 408 | 6496101 | ||
BG748458.1 | - | 7 | 486 | |||
BM822755.1 | - | 8 | 515 | |||
EU339107.1 | - | 4 | 209 | |||
FJ985780.1 | - | 6 | 434 | |||
FJ985781.1 | - | 1 | 40 | |||
M59807.1 | - | 6 | 526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
