siRNA Details
Gene Symbol: CNOT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190847.1 | NP_001177776.1 | 11 | 2617 | ||
NM_001190849.1 | NP_001177778.1 | 12 | 2821 | |||
NM_001190850.1 | NP_001177779.1 | 12 | 2830 | |||
NM_013316.3 | NP_037448.2 | 11 | 2608 | |||
XM_017012235.1 | XP_016867724.1 | 12 | 2691 | |||
GenBank mRNA | AF091094.1 | - | 8 | 1880 | ||
AK074671.1 | - | 10 | 2017 | |||
AK300365.1 | - | 12 | 2787 | |||
U71267.1 | - | 11 | 2567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
