siRNA Details
Gene Symbol: PABPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030979.2 | NP_112241.2 | 1 | 2121 | ||
GenBank mRNA | AF132026.1 | - | 1 | 2083 | ||
BC045608.1 | - | 1 | 2097 | 5273503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
