siRNA Details
Gene Symbol: RECQL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003715.3 | NP_001003715.1 | 3 | 337 | ||
NM_001003716.3 | NP_001003716.1 | 3 | 337 | |||
NM_004259.6 | NP_004250.4 | 3 | 337 | |||
XM_005257818.3 | XP_005257875.1 | 3 | 350 | |||
XM_005257823.3 | XP_005257880.1 | 3 | 349 | |||
XM_011525482.2 | XP_011523784.1 | 2 | 817 | |||
XM_017025343.1 | XP_016880832.1 | 2 | 817 | |||
XM_017025344.1 | XP_016880833.1 | 2 | 817 | |||
GenBank mRNA | AB006533.1 | - | 2 | 177 | ||
AB042823.1 | - | 3 | 337 | |||
AB042824.1 | - | 3 | 337 | |||
AB042825.1 | - | 3 | 337 | |||
AK075084.1 | - | 3 | 328 | |||
AK304168.1 | - | 3 | 329 | |||
BC016911.1 | - | 3 | 513 | |||
DB091647.1 | - | 3 | 333 | |||
EF560729.1 | - | 3 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
