siRNA Details
Gene Symbol: NR3C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000176.2 | NP_000167.1 | 9 | 3809 | ||
NM_001018074.1 | NP_001018084.1 | 9 | 3639 | |||
NM_001018075.1 | NP_001018085.1 | 9 | 3542 | |||
NM_001018076.1 | NP_001018086.1 | 9 | 3435 | |||
NM_001018077.1 | NP_001018087.1 | 9 | 4311 | |||
NM_001024094.1 | NP_001019265.1 | 9 | 3812 | |||
NM_001204258.1 | NP_001191187.1 | 9 | 3809 | |||
NM_001204259.1 | NP_001191188.1 | 9 | 3809 | |||
NM_001204260.1 | NP_001191189.1 | 9 | 3809 | |||
NM_001204261.1 | NP_001191190.1 | 9 | 3809 | |||
NM_001204262.1 | NP_001191191.1 | 9 | 3809 | |||
NM_001204263.1 | NP_001191192.1 | 9 | 3809 | |||
NM_001204264.1 | NP_001191193.1 | 9 | 3809 | |||
XM_011537637.2 | XP_011535939.1 | 8 | 4244 | |||
XM_017009397.1 | XP_016864886.1 | 9 | 3435 | |||
XM_017009398.1 | XP_016864887.1 | 9 | 3734 | |||
GenBank mRNA | AM183262.1 | - | 9 | 3252 | ||
X03225.1 | - | 9 | 3449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
