siRNA Details
Gene Symbol: CHST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004267.4 | NP_004258.2 | 2 | 1925 | ||
GenBank mRNA | AB014679.2 | - | 2 | 1234 | ||
AB014680.2 | - | 2 | 1234 | |||
AB021124.1 | - | 2 | 2370 | |||
AF083066.1 | - | 1 | 1043 | |||
AK023268.1 | - | 2 | 1875 | |||
BC017499.1 | - | 2 | 1606 | |||
BC036930.1 | - | 2 | 1836 | |||
BC068604.1 | - | 2 | 1875 | |||
BC105010.1 | - | 1 | 1005 | 8144013 | ||
BC105012.1 | - | 1 | 1005 | 8144015 | ||
EU668328.1 | - | 1 | 1005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
