siRNA Details
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146099.1 | NP_001139571.1 | 2, 3 | 491 | ||
NM_002498.2 | NP_002489.1 | 2, 3 | 491 | |||
NM_152720.2 | NP_689933.1 | 2, 3 | 510 | |||
NR_027415.2 | 2, 3 | 491 | ||||
GenBank mRNA | AB072828.1 | - | 1, 2 | 160 | ||
AK131359.1 | - | 1, 2 | 1409 | |||
AK290259.1 | - | 2, 3 | 223 | |||
AL833951.1 | - | 2, 3 | 350 | |||
BC019916.1 | - | 2, 3 | 218 | 5000918 | ||
BQ648157.1 | - | 2, 3 | 427 | |||
DA733733.1 | - | 2, 3 | 491 | |||
DB185803.1 | - | 2, 3 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
